Thermo Scientific Chemicals

Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A12691.06
5 g, Each
Thermo Scientific Chemicals

Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A12691.06
5 g, Each
Quantity
Catalog number: A12691.06
also known as A12691-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3849-21-6
IUPAC Name
(E)-(ethyl cyano(hydroxyimino)formate)
Molecular Formula
C5H6N2O3
InChI Key
LCFXLZAXGXOXAP-QPJJXVBHSA-N
SMILES
CCOC(=O)C(=N\O)\C#N
Appearance (Color)
White to pale cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
119.0-134.0?C

Description

This product is a non-explosive alternative to HOBt or HOAt, and allows high peptide coupling rates at low racemization when applied in combination with carbodiimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text