Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals
Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A12691.22
also known as A12691-22
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
Add to cart
Ethyl isonitrosocyanoacetate, 97%, Thermo Scientific Chemicals
Catalog numberA12691.22
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3849-21-6
IUPAC Name(E)-(ethyl cyano(hydroxyimino)formate)
Molecular FormulaC5H6N2O3
InChI KeyLCFXLZAXGXOXAP-QPJJXVBHSA-N
SMILESCCOC(=O)C(=N\O)\C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)119.0-134.0?C
This product is a non-explosive alternative to HOBt or HOAt, and allows high peptide coupling rates at low racemization when applied in combination with carbodiimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only