4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A12693.14
also known as A12693-14
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Catalog numberA12693.14
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS2131-55-7
IUPAC Name1-chloro-4-isothiocyanatobenzene
Molecular FormulaC7H4ClNS
InChI KeyMZZVFXMTZTVUFO-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point41-47?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only