Thermo Scientific Chemicals

1-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12699.14
25 g, Each
Thermo Scientific Chemicals

1-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12699.14
25 g, Each
Quantity
Catalog number: A12699.14
also known as A12699-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
665-66-7
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
<0.5%
Form
Powder or crystalline powder
Identification (FTIR)
Conforms

Description

1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text