1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only
General References:
- A.W. Galbraith; J.S. Oxford; G.C. Schild; G.I. Watson. Protective Effect Of 1-Adamantanamine Hydrochloride On Influenza A2 Infections In The Family Environment. The Lancet. 1969, 294(7629),1026-1028.
- W. L. Davies; R. R. Grunert; R. F. Haff; J. W. McGahen; E. M. Neumayer; M. Paulshock; J. C. Watts; T. R. Wood; E. C. Hermann; C. E. Hoffmann. Antiviral Activity of 1-Adamantanamine (Amantadine). Science . 1964, 144(3620),862-863.