Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.
Solubility
Soluble in water
Notes
Being a mild reducing agent, it gets converted into iodine by oxidizing reagents. Potassium iodide should be handled with gloves since it is a mild irritant.
RUO – Research Use Only
Hajipour, A. R.; Adibi, H. Highly selective iodination of phenols using potassium iodide and benzyltriphenylphosphonium peroxymonosulfate. J. Chem. Res. 2004, 2004 (4), 294-295.
Saatchi, M.; Hosseini, H. S.; Farhad, A. R.; Narimany, T. The effect of various concentrations of iodine potassium iodide on the antimicrobial properties of mineral trioxide aggregate - a pilot study. Dent. Traumatol. 2012, 28, 474-477.
Milczarek, M.; Stepniak, J.; Lewinski, A.; Karbownik-Lewinska, M. “Potassium iodide, but not potassium iodate, as a potential protective agent against oxidative damage to membrane lipids in porcine thyroid.” Thyroid Res. 2013, 6, 10.