Sodium antimonyl L-tartrate, 98+%, Thermo Scientific Chemicals
Sodium antimonyl L-tartrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium antimonyl L-tartrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
Catalog number A12707.0B
also known as A12707-0B
Price (USD)/ Each
381.00
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
381.00
Add to cart
Sodium antimonyl L-tartrate, 98+%, Thermo Scientific Chemicals
Catalog numberA12707.0B
Price (USD)/ Each
381.00
-
Add to cart
Chemical Identifiers
CAS34521-09-0
Molecular FormulaC8H4Na2O12Sb2
InChI KeyUZVPADYOEPQJSP-UHFFFAOYNA-J
SMILES[Na+].[Na+].[Sb+3].[Sb+3].[O-]C(C([O-])C([O-])=O)C([O-])=O.[O-]C(C([O-])C([O-])=O)C([O-])=O
Molecular Weight (g/mol)581.61
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration)≥98%
FormPowder
Assay from Supplier's CofALoss on drying: 0 - 2 wt%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only