4-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals
4-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A12711.30
also known as A12711-30
Price (USD)/ Each
187.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
187.00
Add to cart
4-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA12711.30
Price (USD)/ Each
187.00
-
Add to cart
Chemical Identifiers
CAS556-08-1
IUPAC Name4-acetamidobenzoic acid
Molecular FormulaC9H9NO3
InChI KeyQCXJEYYXVJIFCE-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale gray
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only