Thermo Scientific Chemicals

Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals

Catalog number: A12712.22
100 g, Each
Thermo Scientific Chemicals

Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals

Catalog number: A12712.22
100 g, Each
Quantity
Catalog number: A12712.22
also known as A12712-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-32-7
IUPAC Name
5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular Formula
C10H2O6
InChI Key
ANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
281.0-291.0?C
Appearance (Color)
White to pale cream

Description

Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.

Solubility
Soluble in dimethyl sulfoxide, acetone, chloroform, ethyl ether, n-hexane and benzene.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text