Thermo Scientific Chemicals

Barium hydroxide octahydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12714.0C
10000 g, Each
Thermo Scientific Chemicals

Barium hydroxide octahydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12714.0C
10000 g, Each
Quantity
Catalog number: A12714.0C
also known as A12714-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12230-71-6
IUPAC Name
barium(2+) octahydrate dihydroxide
Molecular Formula
BaH18O10
InChI Key
ZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%
Loss on Drying
42.5-48.5% (200?C/constant weight)
Appearance (Color)
White
Carbonate
<4.0% as BaCO3

Description

Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

Solubility
Soluble in water (72 mg/ml at 20°C), methanol, ethanol (slightly), and water (56 mg/ml at 15°C). Insoluble in acetone (practically)

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text