Procion Blue H-5R
Thermo Scientific Chemicals

Procion Blue H-5R

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12719.14
also known as A12719-14
Price (USD)/ Each
47.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
47.60
Add to cart
Procion Blue H-5R
Catalog numberA12719.14
Price (USD)/ Each
47.60
-
Add to cart
Chemical Identifiers
CAS12236-84-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue or black
Solution Test10mg in100ml water: Clear blue or purple solution
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only