L-Valine, 99%
Thermo Scientific Chemicals

L-Valine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A12720.36
also known as A12720-36
Price (USD)/ Each
650.65
Online exclusive
723.00 
Save 72.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
650.65
Online exclusive
723.00 
Save 72.35 (10%)
Add to cart
L-Valine, 99%
Catalog numberA12720.36
Price (USD)/ Each
650.65
Online exclusive
723.00 
Save 72.35 (10%)
-
Add to cart
Chemical Identifiers
CAS72-18-4
IUPAC Name(2S)-2-amino-3-methylbutanoic acid
Molecular FormulaC5H11NO2
InChI KeyKZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILESCC(C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation+26.8? ± 1.2? (c=5, 4M HCl)
L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in methanol and acetone.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Liang, C.; Huo, Y.; Qi, G.; Wei, X.; Wang, Q.; Chen, S. Enhancement of L-valine production in Bacillus licheniformis by blocking three branched pathways. Biotechnol. Lett. 2015, 37 (6), 1243-1248.
  2. Mahr, R.; Gatgens, C.; Gatgens, J.; Polen, T.; Kalinowski, J.; Frunzke, J. Biosensor-driven adaptive laboratory evolution of L-valine production in Corynebacterium glutamicum. Metab. Eng. 2015, 32, 184-194.