Thermo Scientific Chemicals

tert-Butyldiphenylchlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: A12721.30
250 g, Each
Thermo Scientific Chemicals

tert-Butyldiphenylchlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: A12721.30
250 g, Each
Quantity
Catalog number: A12721.30
also known as A12721-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58479-61-1
IUPAC Name
tert-butyl(chloro)diphenylsilane
Molecular Formula
C16H19ClSi
InChI Key
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥96.0%
Refractive Index
1.5665-1.5700 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow or pink

Description

tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

Solubility
Miscible with chloroform and ethyl acetate. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text