Thermo Scientific Chemicals

Ethylene chlorophosphite, 97%, Thermo Scientific Chemicals

Catalog number: A12724.14
25 g, Each
Thermo Scientific Chemicals

Ethylene chlorophosphite, 97%, Thermo Scientific Chemicals

Catalog number: A12724.14
25 g, Each
Quantity
Catalog number: A12724.14
also known as A12724-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
822-39-9
IUPAC Name
2-chloro-1,3,2-dioxaphospholane
Molecular Formula
C2H4ClO2P
InChI Key
OLSFRDLMFAOSIA-UHFFFAOYSA-N
SMILES
ClP1OCCO1
Form
Liquid
Refractive Index
1.4890-1.4940 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

Notes
Store in cool place. Moisture sensitive. Incompatible with water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text