Thermo Scientific™

4-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12725.03
1 g, Each
Thermo Scientific™

4-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12725.03
1 g, Each
Quantity
Catalog number: A12725.03
also known as A12725-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
825-83-2
IUPAC Name
[4-(trifluoromethyl)phenyl]sulfanide
Molecular Formula
C7H4F3S
InChI Key
WCMLRSZJUIKVCW-UHFFFAOYSA-M
SMILES
FC(F)(F)C1=CC=C([S-])C=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4895-1.4945 @ 20?C

Description

4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text