Thermo Scientific Chemicals

3,5-Dibromobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A12726.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dibromobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A12726.06
5 g, Each
Quantity
Catalog number: A12726.06
also known as A12726-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56908-88-4
IUPAC Name
1,3-dibromo-5-(bromomethyl)benzene
Molecular Formula
C7H5Br3
InChI Key
PWTFRUXTAFBWBW-UHFFFAOYSA-N
SMILES
BrCC1=CC(Br)=CC(Br)=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
92.0-98.0?C
Appearance (Color)
Pale yellow
Assay (GC)
≥98.5%

Description

3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in toluene

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to water, oxidizing agents, bases, alcohols, amines, steel.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text