Thermo Scientific Chemicals

Silicone oil, for oil baths, usable range from -40 to +200°C, Thermo Scientific Chemicals

Catalog number: A12728.0E
2500 g, Each
Thermo Scientific Chemicals

Silicone oil, for oil baths, usable range from -40 to +200°C, Thermo Scientific Chemicals

Catalog number: A12728.0E
2500 g, Each
Quantity
Catalog number: A12728.0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63148-62-9
IUPAC Name
Polydimethylsiloxane
Molecular Formula
(C2H6OSi)n
SMILES
C[Si](C)(-*)O-*
MDL Number
MFCD00132673
Form
Viscous liquid
Identification (FTIR)
Conforms
Viscosity
45-55 mm²/s @ 25?C
Appearance (Color)
Clear colorless
Comment
May produce trace quantities of formaldehyde above 150?C

Description

Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

Solubility
Miscible with aliphatic, aromatic hydrocarbons, alcohol and chlorinated hydrocarbons. Immiscible with water, methanol and ethylene glycol.

Notes
Viscosity at 40cSt. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text