Thermo Scientific Chemicals

Ethyl succinyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A12734.14
25 g, Each
Thermo Scientific Chemicals

Ethyl succinyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A12734.14
25 g, Each
Quantity
Catalog number: A12734.14
also known as A12734-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14794-31-1
IUPAC Name
ethyl 4-chloro-4-oxobutanoate
Molecular Formula
C6H9ClO3
InChI Key
IXZFDJXHLQQSGQ-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC(Cl)=O
Refractive Index
1.4345-1.4395 @ 20°C
Appearance (Color)
Clear colorless to yellow to brown
Assay (Methyl Ester GC)
≥94.0%
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text