Thermo Scientific Chemicals

2-Methylthiophene, 98%, Thermo Scientific Chemicals

Catalog number: A12736.22
100 g, Each
Thermo Scientific Chemicals

2-Methylthiophene, 98%, Thermo Scientific Chemicals

Catalog number: A12736.22
100 g, Each
Quantity
Catalog number: A12736.22
also known as A12736-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-14-3
IUPAC Name
2-methylthiophene
Molecular Formula
C5H6S
InChI Key
XQQBUAPQHNYYRS-UHFFFAOYSA-N
SMILES
CC1=CC=CS1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5185-1.5225 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text