Thermo Scientific Chemicals

Oxalacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12739.06
5 g, Each
Thermo Scientific Chemicals

Oxalacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12739.06
5 g, Each
Quantity
Catalog number: A12739.06
also known as A12739-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
328-42-7
IUPAC Name
2-oxobutanedioic acid
Molecular Formula
C4H4O5
InChI Key
KHPXUQMNIQBQEV-UHFFFAOYSA-N
SMILES
OC(=O)CC(=O)C(O)=O
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.

Solubility
Soluble in water (0.1 g/ml), ether, ethanol, acetone, and acetone ethyl acetate.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from heat and sunlight.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text