Thermo Scientific Chemicals

2-n-Butylthiophene, 98+%, Thermo Scientific Chemicals

Catalog number: A12740.14
25 g, Each
Thermo Scientific Chemicals

2-n-Butylthiophene, 98+%, Thermo Scientific Chemicals

Catalog number: A12740.14
25 g, Each
Quantity
Catalog number: A12740.14
also known as A12740-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1455-20-5
IUPAC Name
2-butylthiophene
Molecular Formula
C8H12S
InChI Key
MNDZHERKKXUTOE-UHFFFAOYSA-N
SMILES
CCCCC1=CC=CS1
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4995-1.5035 @ 20°C

Description

2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

Solubility
Soluble in acetone and DMSO.

Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text