Thermo Scientific Chemicals

Lead(IV) oxide, 97%, Thermo Scientific Chemicals

Catalog number: A12742.36
500 g, Each
Thermo Scientific Chemicals

Lead(IV) oxide, 97%, Thermo Scientific Chemicals

Catalog number: A12742.36
500 g, Each
Quantity
Catalog number: A12742.36
also known as A12742-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-60-0
IUPAC Name
plumbanedione
Molecular Formula
O2Pb
InChI Key
YADSGOSSYOOKMP-UHFFFAOYSA-N
SMILES
O=[Pb]=O
Appearance (Color)
Brown or black
Assay from Supplier's CofA
≥96.0 to ≤104.0%
Form
powder

Description

Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

Solubility
Insoluble in water. Soluble in dil.HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text