trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A12745.14
also known as A12745-14
Price (USD)/ Each
343.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
343.00
Add to cart
trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
Catalog numberA12745.14
Price (USD)/ Each
343.00
-
Add to cart
Chemical Identifiers
CAS5576-97-6
IUPAC Name1-methoxy-4-(2-nitroethenyl)benzene
Molecular FormulaC9H9NO3
InChI KeyJKQUXSHVQGBODD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C[N+]([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)84.5-90.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only