Thermo Scientific Chemicals

Phenylselenenyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12751.06
5 g, Each
Thermo Scientific Chemicals

Phenylselenenyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12751.06
5 g, Each
Quantity
Catalog number: A12751.06
also known as A12751-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5707-04-0
IUPAC Name
(phenylselanyl)chlorane
Molecular Formula
C6H5ClSe
InChI Key
WJCXADMLESSGRI-UHFFFAOYSA-N
SMILES
Cl[Se]C1=CC=CC=C1
Appearance (Color)
Orange to red to dark red
Form
Crystals or Crystalline powder and/or chunks
Solution Test
(5% w/v in ethanol) : Clear solution
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with strong bases, strong oxidizing agents and strong alkalis.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text