Thermo Scientific Chemicals

Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A12754.0B
1000 g, Each
Thermo Scientific Chemicals

Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A12754.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12754.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-09-7
IUPAC Name
ethyl 4-aminobenzoate
Molecular Formula
C9H11NO2
InChI Key
BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(N)C=C1
Appearance (Color)
White to pale cream
Melting Point (clear melt)
87.0-94.0?C
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.

Solubility
Soluble in ethanol, chloroform, ethyl ether and dilute acids. Sparingly soluble in water

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text