6-Chloro-1,3-dimethyluracil, 97%, Thermo Scientific Chemicals
6-Chloro-1,3-dimethyluracil, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloro-1,3-dimethyluracil, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A12755.06
also known as A12755-06
Price (USD)/ Each
95.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
95.30
Add to cart
6-Chloro-1,3-dimethyluracil, 97%, Thermo Scientific Chemicals
Catalog numberA12755.06
Price (USD)/ Each
95.30
-
Add to cart
Chemical Identifiers
CAS6972-27-6
IUPAC Name6-chloro-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC6H7ClN2O2
InChI KeyVATQPUHLFQHDBD-UHFFFAOYSA-N
SMILESCN1C(Cl)=CC(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)107.0-116.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only