Thermo Scientific Chemicals

4-Chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12757.0B
1000 g, Each
Thermo Scientific Chemicals

4-Chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12757.0B
1000 g, Each
Quantity
Catalog number: A12757.0B
also known as A12757-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-88-1
IUPAC Name
4-chlorobenzaldehyde
Molecular Formula
C7H5ClO
InChI Key
AVPYQKSLYISFPO-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=O)C=C1
Assay (GC)
≥97.5%
Appearance (Color)
White to pale cream or pale yellow
Form
Crystals or powder or crystalline powder or fused solid or chunks
Free acid (titration)
≤2.5%
Identification (FTIR)
Conforms

Description

4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

Solubility
Soluble in water (0.94g/L).

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text