Thermo Scientific Chemicals

DL-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12758.14
25 g, Each
Thermo Scientific Chemicals

DL-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12758.14
25 g, Each
Quantity
Catalog number: A12758.14
also known as A12758-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5619-07-8
IUPAC Name
methyl 2-amino-3-phenylpropanoate hydrochloride
Molecular Formula
C10H14ClNO2
InChI Key
SWVMLNPDTIFDDY-UHFFFAOYNA-N
SMILES
Cl.COC(=O)C(N)CC1=CC=CC=C1
Appearance (Color)
White
Form
Crystalline powder
Assay (Titration ex Chloride)
>97.5 to <102.5%
Water Content (Karl Fischer Titration)
<0.5%

Description

DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text