4-Isopropoxybenzaldehyde, 97%, Thermo Scientific Chemicals
4-Isopropoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A12760.06
also known as A12760-06
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
4-Isopropoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberA12760.06
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS18962-05-5
IUPAC Name4-(propan-2-yloxy)benzaldehyde
Molecular FormulaC10H12O2
InChI KeyWDANSDASCKBVKH-UHFFFAOYSA-N
SMILESCC(C)OC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5440-1.5490 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only