2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water (less).
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Yong Guo; Bingjun Din; Yongwen Liuc; Xijun Chang; Shuangming Meng; Maozhong Tian. Preconcentration of trace metals with 2-(methylthio)aniline-functionalized XAD-2 and their determination by flame atomic absorption spectrometry. Analytica Chimica Acta. 2004, 504(2), 319-324.
- Christopher G. Hamaker; Oksana S. Maryashina; Dana K. Daley; Andrew L. Wadler. Synthesis and Crystal Structures of Two Schiff Bases of 2-(Methylthio)aniline with Halogenated Salicylaldehydes. Journal of Chemical Crystallography. 2010, 40(1), 34-39.