Thermo Scientific Chemicals

2-(Methylthio)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A12762.22
100 g, Each
Thermo Scientific Chemicals

2-(Methylthio)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A12762.22
100 g, Each
Quantity
Catalog number: A12762.22
also known as A12762-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2987-53-3
IUPAC Name
2-(methylsulfanyl)aniline
Molecular Formula
C7H9NS
InChI Key
WBRPQQSADOCKCH-UHFFFAOYSA-N
SMILES
CSC1=CC=CC=C1N
Form
Liquid
Refractive Index
1.6220-1.6260 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear yellow to red to brown

Description

2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (less).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text