Benzimidazole, 99%, Thermo Scientific Chemicals
Benzimidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzimidazole, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
50g
Catalog number A12763.18
also known as A12763-18
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.80 
Save 4.15 (13%)
-
Add to cart
Quantity:
50g
Request bulk or custom format
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.80 
Save 4.15 (13%)
Add to cart
Benzimidazole, 99%, Thermo Scientific Chemicals
Catalog numberA12763.18
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
30.80 
Save 4.15 (13%)
-
Add to cart
Chemical Identifiers
CAS51-17-2
IUPAC Name1H-1,3-benzodiazole
Molecular FormulaC7H6N2
InChI KeyHYZJCKYKOHLVJF-UHFFFAOYSA-N
SMILESN1C=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Melting Point (clear melt)169-174?C
View more
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Chaudhry, U.; Redman, E. M.; Raman, M.; Gilleard, J. S. Genetic evidence for the spread of a benzimidazole resistance mutation across southern India from a single origin in the parasitic nematode Haemonchus contortus. Int J Parasitol 2015, 45 (11), 721-728.

Dam, C. S.; Henarejos, S. A. P.; Tsolakou, T.; Segato, C. A.; Gammelgaard, B.; Yellol, G. S.; Ruiz, J.; Lambert, I. H.; Sturup, S. In vitro characterization of a novel C,N-cyclometalated benzimidazole Ru(II) arene complex: stability, intracellular distribution and binding, effects on organic osmolyte homeostasis and induction of apoptosis. Metallomics 2015, 7 (5), 885-895.