Thermo Scientific Chemicals

1,2-Dibromoethane, 99%, Thermo Scientific Chemicals

Catalog number: A12766.0B
1000 g, Each
Thermo Scientific Chemicals

1,2-Dibromoethane, 99%, Thermo Scientific Chemicals

Catalog number: A12766.0B
1000 g, Each
Quantity
Catalog number: A12766.0B
also known as A12766-0B
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-93-4
IUPAC Name
1,2-dibromoethane
Molecular Formula
C2H4Br2
InChI Key
PAAZPARNPHGIKF-UHFFFAOYSA-N
SMILES
BrCCBr
Assay from Supplier's CofA
≥97.5% (GC, US-sourced material)
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5% (UK-sourced material)
Comment
Material sourced in UK and US

Description

1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

Solubility
Miscible with alcohol, acetone, benzene, diethyl ether and ethyl acetate. Slightly miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text