1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.
Solubility
Sparingly Soluble (4.0E-3 g/L) (25°C).
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Wang X, et al. Synthesis and characterization of ABC-type star and linear block copolymers of styrene, isoprene, and 1, 3-cyclohexadiene. Macromolecules. 2006,39(20), 6898-6904.
Shigeaki Nakazawa et. al. Organic polyanionic high-spin molecular clusters: topological-symmetry controlled models for organic ferromagnetic metals. Physical Chemistry Chemical Physics. 2011,13(4), 1424-1433.