Diethyl ketomalonate, 95%, Thermo Scientific Chemicals
Diethyl ketomalonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl ketomalonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A12771.22
also known as A12771-22
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
Add to cart
Diethyl ketomalonate, 95%, Thermo Scientific Chemicals
Catalog numberA12771.22
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
-
Add to cart
Chemical Identifiers
CAS609-09-6
IUPAC Name1,3-diethyl 2-oxopropanedioate
Molecular FormulaC7H10O5
InChI KeyDBKKFIIYQGGHJO-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
Refractive Index1.4145-1.4205 @ 20?C
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

Solubility
Fully miscible with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

General References:

  1. D. Basavaiah; V. V. L. Gownswari. Diethyl Ketomalonate: A Fast Reacting Substrate for Baylis-Hillman Reaction. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1989,(13-14), 6898-6904.
  2. M. Sugawara; M.M. Baizer. Electrogenerated bases VII. Novel syntheses of ethyl glyoxalate and diethyl ketomalonate via electrogenerated superoxide. Tetrahedron Letters. 1983,24(22), 2223-2226.
  3. The carbonyl group behaves as a heterodienophile towards simple dienes. Curtius degradation of the gem-diester to carbonyl completes a synthesis of ß-unsaturated δ-lactones: Rocz. Chem., 38, 707 (1964); J. Am. Chem. Soc., 97, 6892 (1975); J. Org. Chem., 42, 4095 (1977):
  4. For asymmetric Diels-Alder reaction, see: J. Chem. Soc., Chem. Commun., 676 (1987). For addition to aza-dienes, see: Tetrahedron Lett, 30, 2685 (1989).
  5. Ene-addition to alkenes occurs at the less-substituted carbon. Oxidative degradation leads to ß-unsaturated carboxylic acids: J. Am. Chem. Soc., 102, 2473 (1980); 106, 3797 (1984):
  6. Thus, the reagent could be regarded as a CO2 equivalent in both the above sequences.
  7. Undergoes the Baylis-Hillman reaction in which a carbanionic species, generated by the reversible addition of 1,4-Diazabicyclo[2.2.2]octane, A14003, to a Michael acceptor, adds to the carbonyl group: Synth. Commun., 19, 2461 (1989).
  8. Reacts with activated arenes under Friedel-Crafts conditions, or with aryllithium or Grignard derivatives, to give aryl hydroxymalonates, hydrolysis and decarboxylation of which gives substituted mandelic acids: Org. Synth. Coll., 3, 326 (1955). Alternatively, they may be transformed reductively to arylmalonates and hence to ester enolates: J. Org. Chem., 47, 4692 (1982).