Thermo Scientific Chemicals

Di-tert-butyl malonate, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A12774.09
10 g, Each
Thermo Scientific Chemicals

Di-tert-butyl malonate, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A12774.09
10 g, Each
Quantity
Catalog number: A12774.09
also known as A12774-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-16-2
IUPAC Name
1,3-di-tert-butyl propanedioate
Molecular Formula
C11H20O4
InChI Key
CLPHAYNBNTVRDI-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
Appearance (Color)
Clear, colorless
Assay (GC)
>98.0%
Form
Liquid
Refractive Index
1.4170-1.4210

Description

Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

Solubility
It is hardly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text