Thermo Scientific™

1,2-Dichloroethane, 99+%, Thermo Scientific Chemicals

Catalog number: A12775.AP
500 mL, Each
Thermo Scientific™

1,2-Dichloroethane, 99+%, Thermo Scientific Chemicals

Catalog number: A12775.AP
500 mL, Each
Quantity
Catalog number: A12775.AP
also known as A12775-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-06-2
IUPAC Name
1,2-dichloroethane
Molecular Formula
C2H4Cl2
InChI Key
WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES
ClCCCl
Color (APHA)
≤10 (U.S. sourced Material)
Assay from Supplier's CofA
≥99.0% (U.S. sourced Material)
Refractive Index
1.4438-1.4458 @20°C (non-U.S. sourced material)
Form
Liquid
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

Solubility
Miscible with methanol, diethyl ether, n-octanol and acetone. Very slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, bases and alkali metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text