3,5-Dibromo-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
3,5-Dibromo-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dibromo-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12780.14
also known as A12780-14
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
3,5-Dibromo-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA12780.14
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS2973-77-5
IUPAC Name3,5-dibromo-4-hydroxybenzaldehyde
Molecular FormulaC7H4Br2O2
InChI KeySXRHGLQCOLNZPT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(C=O)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or crystals or needles
Identification (FTIR)Conforms
Melting Point (clear melt)178-186?C
Assay (HPLC)≥96.0%
Appearance (Color)White to cream to pink
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only