Thermo Scientific Chemicals

Difluoroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12787.06
5 g, Each
Thermo Scientific Chemicals

Difluoroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12787.06
5 g, Each
Quantity
Catalog number: A12787.06
also known as A12787-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
381-73-7
IUPAC Name
2,2-difluoroacetic acid
Molecular Formula
C2H2F2O2
InChI Key
PBWZKZYHONABLN-UHFFFAOYSA-N
SMILES
OC(=O)C(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Refractive Index
1.3410-1.3450 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text