4'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific Chemicals
4'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12789.14
also known as A12789-14
Price (USD)/ Each
256.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
256.00
Add to cart
4'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific Chemicals
Catalog numberA12789.14
Price (USD)/ Each
256.00
-
Add to cart
Chemical Identifiers
CAS349-97-3
IUPAC NameN-[4-(trifluoromethyl)phenyl]acetamide
Molecular FormulaC9H8F3NO
InChI KeyDFDHFECLWHHELH-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)149.0-155.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only