1-Heptanol, 99%, Thermo Scientific Chemicals
1-Heptanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Heptanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
500 mL
Catalog number A12793.0F
also known as A12793-0F
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
Add to cart
1-Heptanol, 99%, Thermo Scientific Chemicals
Catalog numberA12793.0F
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS111-70-6
IUPAC Nameheptan-1-ol
Molecular FormulaC7H16O
InChI KeyBBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILESCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Refractive Index1.4230-1.4260 @ 20?C
Assay (GC)≥98.5%
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

General References:

  1. NV Sastry.; MK Valand. Volumetric behaviour of acrylic esters (methyl-, ethyl-, and butyl acrylate)+ 1-alcohol (heptanol, octanol, decanol and dodecanol) at 298.15 K and 308.15 K. Physics and chemistry of liquids. 200038, (1) , 61-72.
  2. AS Al-Jimaz.; JA Al-Kandary.; AHM Abdul-Latif . Densities and viscosities for binary mixtures of phenetole with 1-pentanol, 1-hexanol, 1-heptanol, 1-octanol, 1-nonanol, and 1-decanol at different temperatures. Fluid phase equilibria. 2004218, (2) , 247-260.