Thermo Scientific Chemicals

1-Heptanol, 99%, Thermo Scientific Chemicals

Catalog number: A12793.AP
500 mL, Each
Thermo Scientific Chemicals

1-Heptanol, 99%, Thermo Scientific Chemicals

Catalog number: A12793.AP
500 mL, Each
Quantity
Catalog number: A12793.AP
also known as A12793-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-70-6
IUPAC Name
heptan-1-ol
Molecular Formula
C7H16O
InChI Key
BBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILES
CCCCCCCO
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.4230-1.4260 @ 20?C
Assay (GC)
≥98.5%

Description

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text