Thermo Scientific Chemicals

5-Fluoroindole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12794.14
25 g, Each
Thermo Scientific Chemicals

5-Fluoroindole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12794.14
25 g, Each
Quantity
Catalog number: A12794.14
also known as A12794-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
399-76-8
IUPAC Name
5-fluoro-1H-indole-2-carboxylate
Molecular Formula
C9H5FNO2
InChI Key
WTXBRZCVLDTWLP-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC2=CC(F)=CC=C2N1
Appearance (Color)
Cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%
Form
Powder

Description

5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text