Thermo Scientific Chemicals

2,6-Dimethyl-4-pyrone, 99%, Thermo Scientific Chemicals

Catalog number: A12799.09
10 g, Each
Thermo Scientific Chemicals

2,6-Dimethyl-4-pyrone, 99%, Thermo Scientific Chemicals

Catalog number: A12799.09
10 g, Each
Quantity
Catalog number: A12799.09
also known as A12799-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1004-36-0
IUPAC Name
2,6-dimethyl-4H-pyran-4-one
Molecular Formula
C7H8O2
InChI Key
VSYFZULSKMFUJJ-UHFFFAOYSA-N
SMILES
CC1=CC(=O)C=C(C)O1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
131.0-137.0?C
Assay (GC)
≥98.5%
Appearance (Color)
White to cream to pale brown

Description

2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text