Thermo Scientific Chemicals

Brilliant Green

Catalog number: A12801.14
25 g, Each
Thermo Scientific Chemicals

Brilliant Green

Catalog number: A12801.14
25 g, Each
Quantity
Catalog number: A12801.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
633-03-4
IUPAC Name
bis[4-(diethylamino)phenyl](phenyl)methylium hydrogen sulfate
Molecular Formula
C27H34N2O4S
InChI Key
NNBFNNNWANBMTI-UHFFFAOYSA-M
SMILES
OS([O-])(=O)=O.CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(CC)CC
Form
Crystals or crystalline powder
Comment
Material sourced in the U.S. and in other countries
lambda-max (UV-Vis)
625-631 nm (50:50 IMS:water, non-U.S. sourced material)
Comment
Dye Content: ≥96.0% (U.S. sourced material)
Specific Absorbance (E 1%/1cm)
≥1900 at lambda max (1%, 1 cm, 50:50 Ethanol:Water, non-U.S. sourced material)

Description

Brilliant Green a stain used in bacteriological media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Green a stain used in bacteriological media.

Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).

Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text