Thermo Scientific Chemicals

Methyl 3-aminothiophene-2-carboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: A12802.14
25 g, Each
Thermo Scientific Chemicals

Methyl 3-aminothiophene-2-carboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: A12802.14
25 g, Each
Quantity
Catalog number: A12802.14
also known as A12802-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22288-78-4
IUPAC Name
methyl 3-aminothiophene-2-carboxylate
Molecular Formula
C6H7NO2S
InChI Key
TWEQNZZOOFKOER-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(N)C=CS1
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Appearance (Color)
Cream to pale brown
Melting Point (clear melt)
63.0-69.0?C

Description

Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text