3-Thiopheneacetonitrile, 98%, Thermo Scientific Chemicals
3-Thiopheneacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Thiopheneacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12805.14
also known as A12805-14
Price (USD)/ Each
330.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
330.00
Add to cart
3-Thiopheneacetonitrile, 98%, Thermo Scientific Chemicals
Catalog numberA12805.14
Price (USD)/ Each
330.00
-
Add to cart
Chemical Identifiers
CAS13781-53-8
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Clear, colourless to brown
Refractive Index1.5410-.15450
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only