Thermo Scientific Chemicals

Dimethylglyoxime, 99%, Thermo Scientific Chemicals

Catalog number: A12810.0E
2500 g, Each
Thermo Scientific Chemicals

Dimethylglyoxime, 99%, Thermo Scientific Chemicals

Catalog number: A12810.0E
2500 g, Each
Quantity
Catalog number: A12810.0E
also known as A12810-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-45-4
IUPAC Name
(E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
Molecular Formula
C4H8N2O2
InChI Key
JGUQDUKBUKFFRO-GGWOSOGESA-N
SMILES
C\C(=N/O)\C(\C)=N\O
Form
Powder
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥98.5%

Description

Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.

Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text