Thermo Scientific Chemicals

4-n-Butylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A12812.14
25 g, Each
Thermo Scientific Chemicals

4-n-Butylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A12812.14
25 g, Each
Quantity
Catalog number: A12812.14
also known as A12812-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1638-22-8
IUPAC Name
4-butylphenol
Molecular Formula
C10H14O
InChI Key
CYYZDBDROVLTJU-UHFFFAOYSA-N
SMILES
CCCCC1=CC=C(O)C=C1
Form
Liquid
Refractive Index
1.5150-1.5190 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

4-n-Butylphenol is used as an Intermediates of liquid Crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Butylphenol is used as an Intermediates of liquid Crystals.

Solubility
Not miscible with water.

Notes
When not in use, tightly seal the container and store in a dry, cool place. Avoid excessive heat and light. Incompatible to oxidizing agents. Stable if stored under proper conditions. Keep container dry. Keep away from heat. Mechanical exhaust required.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text