Thermo Scientific Chemicals

2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A12820.36
500 g, Each
Thermo Scientific Chemicals

2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A12820.36
500 g, Each
Quantity
Catalog number: A12820.36
also known as A12820-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14338-32-0
IUPAC Name
2-chloro-1-methylpyridin-1-ium iodide
Molecular Formula
C6H7ClIN
InChI Key
ABFPKTQEQNICFT-UHFFFAOYSA-M
SMILES
[I-].C[N+]1=CC=CC=C1Cl
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms
Assay (Titration ex Iodide)
≥96.0 to ≤104.0%
Appearance (Color)
Yellow

Description

2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

Solubility
Soluble in Methanol (50 mg/ml). Insoluble in water.

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text